Learn More
Thermo Scientific™ CaptureSelect™ IX Select Leakage ELISA
Thermo Scientific™ CaptureSelect™ IXSelect Leakage ELISA is designed to detect possible leached ligand of human Factor IX purified using IXSelect affinity resin (Cytiva).
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
810300001
![]() |
Thermo Scientific™
810300001 |
N/A
|
|
||||||
810300010
![]() |
Thermo Scientific™
810300010 |
N/A
|
N/A | ||||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
CaptureSelect IXSelect Leakage ELISA is designed to detect possible leached ligand of human Factor IX purified using IXSelect affinity resin (Cytiva). Even when covalently attached, small amounts of the IXSelect affinity ligand can leach off of the chromatography support and co-elute with the purified product.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect IXSelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.