missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG3 (Human) Conjugate
GREENER_CHOICE

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG3 (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of the human IgG3 subclass, most likely the hinge region.

$1,514.85 - $5,335.33

Specifications

Product Type Biotin Anti-IgG3 Conjugate
Conjugate Biotin and Other Haptens
Product Line CaptureSelect
Content And Storage Store in freezer (-5°C to -30°C).
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
7103042100
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
7103042100
100 μg
Each for $1,514.85
Only null left
 
7103042500
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
7103042500
500 μg
Each for $5,335.33
Only null left
 
Description

Description

CaptureSelect Biotin Anti-IgG3 (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of the human IgG3 subclass, most likely the hinge region. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-IgG3 (Hu) Conjugate allows you to:

  • Detect, quantitate, and characterize human IgG3 antibodies
  • Avoid cross-binding with other human IgG subclasses like IgG1, 2 and 4 and isotypes like IgM and IgA including IgG from bovine sources such as FCS

Applications for CaptureSelect Biotin Anti-IgG3 (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Specifications

Biotin Anti-IgG3 Conjugate
Biotin and Other Haptens
CaptureSelect
Store in freezer (-5°C to -30°C).
Videos
SDS
Documents

Documents