missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-FIX Conjugate
GREENER_CHOICE

Catalog No. 7103002100
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7103002100 100 μg
7103002500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7103002100 Supplier Thermo Scientific™ Supplier No. 7103002100

Thermo Scientific™ CaptureSelect™ Biotin Anti-FIX Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and plasma derived human Factor IX (FIX).

CaptureSelect Biotin Anti-FIX Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and plasma derived human Factor IX (FIX). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-FIX Conjugate allows you to:

  • Detect, quantitate, and characterize recombinant and plasma derived human FIX

Applications for CaptureSelect Biotin Anti-FIX Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Product Type Biotin Anti-FIX Conjugate
Conjugate Biotin and Other Haptens
Quantity 100 μg
Product Line CaptureSelect
Content And Storage Store in freezer (-5°C to -30°C).