missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-Factor X Conjugate
GREENER_CHOICE

Catalog No. 7103702500
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7103702500 500 μg
7103702100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7103702500 Supplier Thermo Scientific™ Supplier No. 7103702500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ CaptureSelect™ Biotin Anti-Factor X Conjugate consists of a 13-kDa recombinant single domain anti-Factor X antibody fragment chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the antibody fragment.

CaptureSelect Biotin Anti-Factor X Conjugate consists of a 13-kDa recombinant single domain anti-Factor X antibody fragment chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the antibody fragment when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize the human coagulation Factor X and Factor Xa.

The antibody fragment specifically binds to human Factor X and its activated form Factor Xa from plasma and recombinant sources. It is highly selective for human Factor X and Xa with no affinity for other human plasma proteins. The ligand binds in a calcium dependent manner, so full release of the bound Factor X or Factor Xa molecule is achieved by addition of chelating agents like EDTA. Suitable binding and target-release conditions are listed below:

  • Binding condition: 50 mM Tris, 150 mM NaCl, 5 mM CaCl2 pH 7
  • Release condition: 50 mM Tris, 150 mM NaCl, 5 mM EDTA pH 7

Applications for CaptureSelect Biotin Anti-Factor X Conjugate include ELISA, immunoprecipitation with streptavidin-coupled Dynabeads, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Coagulation Factor X is a vitamin K-dependent, liver-produced serine protease that serves as the first enzyme in the coagulation cascade to form fibrin. It is a two-chain glycoprotein with a molecular weight of approximately 59 kDa. While Factor X normally circulates in the plasma as an inactive molecule, the activation of Factor X is involved in both the intrinsic and extrinsic coagulation pathways.

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Quantity 500 μg
Product Type Conjugate
Product Line CaptureSelect
Content And Storage Store in freezer (-5°C to -30°C).
Shipping Condition Dry Ice
Label Type Biotin & Analogs