missing translation for 'onlineSavingsMsg'
Learn More

Captan 98.0+%, TCI America™

Catalog No. C20595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20595G 5 g
C205925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20595G Supplier TCI America Supplier No. C20595G
Only null left

Chemical Identifiers

CAS 133-06-2
Molecular Formula C9H8Cl3NO2S
Molecular Weight (g/mol) 300.578
MDL Number MFCD00041811
InChI Key LDVVMCZRFWMZSG-UHFFFAOYSA-N
Synonym captan, amercide, orthocide, vondcaptan, captaf, captane, captex, kaptan, malipur, merpan
PubChem CID 8606
ChEBI CHEBI:34608
IUPAC Name 2-(trichloromethylsulfanyl)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
SMILES C1C=CCC2C1C(=O)N(C2=O)SC(Cl)(Cl)Cl

Specifications

Melting Point 175°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 300.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Captan
EINECSNumber (9)-0934
RTECSNumber GW5075000
TSCA Yes