missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals CAPSO, 99%
CAS: 73463-39-5 | C9H19NO4S | 237.3 g/mol
$212.92 - $212.92
Chemical Identifiers
| CAS | 73463-39-5 |
|---|---|
| Molecular Formula | C9H19NO4S |
| Molecular Weight (g/mol) | 237.3 |
| InChI Key | INEWUCPYEUEQTN-UHFFFAOYSA-N |
| Synonym | capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid |
| PubChem CID | 2733480 |
| IUPAC Name | 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid |
| SMILES | C1CCC(CC1)NCC(CS(=O)(=O)O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC327741000
|
Thermo Scientific Chemicals
327741000 |
100 g | Glass bottle |
Each for $212.92
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
| 73463-39-5 | |
| 237.3 | |
| capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid | |
| 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid |
| C9H19NO4S | |
| INEWUCPYEUEQTN-UHFFFAOYSA-N | |
| 2733480 | |
| C1CCC(CC1)NCC(CS(=O)(=O)O)O |
Specifications
| 73463-39-5 | |
| 263.0°C to 271.0°C | |
| 5 to 6.5 (1% aq. soln. at 25°C) | |
| 99% | |
| C9H19NO4S | |
| capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid | |
| INEWUCPYEUEQTN-UHFFFAOYSA-N | |
| 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid | |
| 2733480 | |
| 99% | |
| CAPSO |
| (0.1M aq. soln.), 0.03 max. at 280nm, (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm | |
| White | |
| Authentic | |
| Glass bottle | |
| 100 g | |
| (0.1M in water) Clear colorless solution | |
| C1CCC(CC1)NCC(CS(=O)(=O)O)O | |
| 237.3 | |
| 237.3 | |
| Powder |
RUO – Research Use Only