Learn More
Thermo Scientific Chemicals CAPSO, 98%
CAS: 73463-39-5 | C9H19NO4S | 237.314 g/mol
Supplier: Thermo Scientific Chemicals B2130536
Description
CAPSO is used in various biological applications such as electrophoresis. Specific applications include buffers for protein separations and diagnostic reagent production. Also used as biological buffer in the following studies, to maintain pH 9 during water leach test for the analysis of high-carbon coal fly ash-soil mixtures, chemiluminescent in situ hybridization using soybean peroxidase-labeled peptide nucleic acid (PNA) probes, preparation of planar bilayer lipid membranes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCAPSO is used in various biological applications such as electrophoresis. Specific applications include buffers for protein separations and diagnostic reagent production. Also used as biological buffer in the following studies, to maintain pH 9 during water leach test for the analysis of high-carbon coal fly ash-soil mixtures, chemiluminescent in situ hybridization using soybean peroxidase-labeled peptide nucleic acid (PNA) probes, preparation of planar bilayer lipid membranes.
Solubility
Soluble in water (10 g+ 90 ml).
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| CAPSO | |
| 270°C to 274°C | |
| MFCD00041778 | |
| Hygroscopic | |
| Soluble in water (10 g+90 ml). | |
| C1CCC(CC1)NCC(CS(=O)(=O)O)O | |
| 237.314 | |
| 237.32 |
| 73463-39-5 | |
| C9H19NO4S | |
| 500 g | |
| capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid | |
| INEWUCPYEUEQTN-UHFFFAOYSA-N | |
| 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid | |
| 2733480 | |
| 98% |
Chemical Identifiers
| 73463-39-5 | |
| 237.314 | |
| INEWUCPYEUEQTN-UHFFFAOYSA-N | |
| 2733480 | |
| C1CCC(CC1)NCC(CS(=O)(=O)O)O |
| C9H19NO4S | |
| MFCD00041778 | |
| capso, 3-cyclohexylamino-2-hydroxy-1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxypropane-1-sulfonic acid, capso free acid, 1-propanesulfonic acid, 3-cyclohexylamino-2-hydroxy, capso compound, 3-cyclohexylamino-2-hydroxy-1-propanesuhicic acid, 3-cyclohexylamino-2-hydroxy-1-propanesulphonic acid, 3-cyclohexylamino-2-hydroxypropanesulfonic acid | |
| 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only