missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ CapSeal Bullet™ Capillary Ferrule, General Purpose, 0.32 mm Column I.D.
Specifications
Specifications
| Diameter (Metric) Inner | 0.5 mm |
| Packaging Quantity | 48/Pk. |
| Description | Ferrule Type: General Purpose |
CapSeal Bullet is a registered trademark of Sigma-Aldrich Co. LLC