missing translation for 'onlineSavingsMsg'
Learn More

CAPS, MP Biomedicals

$125.56 - $125.56

Chemical Identifiers

CAS 1135-40-6
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10143580
View Documents
MP Biomedicals Inc
0210143580
100g
Each for $125.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1135-40-6

Specifications

1135-40-6
100g
CAPS
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'