Learn More
Microsolv Technology Caps For 40Ml Vials Sil/Pt W
Supplier: Microsolv Technology Corporation 9704005CW
Caps, Screw Top, with White Silicone Rubber / White PTFE Septa, fitted in knurled polypropylene, White Caps. For use with 24-414mm threaded EPA Style vials. BASIK Brand. 100/PK.