missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals CAPS, 99%

Catalog No. p-7023464
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1703714 25 g
AAA1703722 100 g
AAA1703736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1703714 Supplier Thermo Scientific Chemicals Supplier No. A1703714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1135-40-6 | C9H19NO3S | 221.32 g/mol

Used to formulate CAPS buffer, a zwitterionic buffer useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electro transfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI > 8.5. and minimal reactivity with enzymes or proteins, minimal salt effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to formulate CAPS buffer, a zwitterionic buffer useful in the range of pH 7.9-11.1. CAPS buffer is widely used in Western and immunoblotting experiments as well as protein sequencing and identification. Used in the electro transfer of proteins to PVDF (sc-3723) or nitrocellulose membranes (sc-3718, sc-3724). The high pH of this buffer makes it useful for the transfer of proteins with a pI>8.5. and minimal reactivity with enzymes or proteins, minimal salt effects.

Solubility
Highly water soluble (100 g/L) at 20°C, and methanol (1%).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
Synonym caps, 3-cyclohexylaminopropanesulfonic acid, n-cyclohexyl-3-aminopropanesulfonic acid, 3-cyclohexylamino-1-propanesulfonic acid, 1-propanesulfonic acid, 3-cyclohexylamino, 3-cyclohexylamino propane-1-sulfonic acid, unii-4w981o1lxp, 3-cyclohexylamino propanesulfonic acid, 3-cyclohexylaminopropane-1-sulphonic acid, 3-cyclohexyl-1-propylsulfonic acid
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Melting Point 324°C
Odor Odorless
Quantity 25 g
Beilstein 2835588
Merck Index 14,1767
Solubility Information Highly water soluble (100g/L) at 20°C,and methanol (1%).
Formula Weight 221.32
Percent Purity 99%
Chemical Name or Material CAPS
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-492-1
RTECSNumber TZ6395000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only