missing translation for 'onlineSavingsMsg'
Learn More

epsilon-Caprolactone monomer, 99%

Catalog No. p-3735924
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173442500 250 g Glass bottle
AC173440010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173442500 Supplier Thermo Scientific Chemicals Supplier No. 173442500
Only null left

CAS: 502-44-3 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 502-44-3
MDL Number MFCD00003267
InChI Key PAPBSGBWRJIAAV-UHFFFAOYSA-N
Synonym 6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane
PubChem CID 10401
ChEBI CHEBI:17915
IUPAC Name oxepan-2-one
SMILES C1CCC(=O)OCC1

Specifications

Melting Point -1°C
Color Colorless
Density 1.0300g/mL
Boiling Point 235°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H10O2
Refractive Index 1.4620 to 1.464
Quantity 250 g
Beilstein 17,II,29
Specific Gravity 1.03
Solubility Information Solubility in water: >1000mg/l (20°C). Other solubilities: soluble in ethanol,acetone,diethyl ether
Molecular Weight (g/mol) 114.14
Formula Weight 114.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material ε-Caprolactone monomer
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 207-938-1

RUO – Research Use Only