missing translation for 'onlineSavingsMsg'
Learn More

3M™ DBI-SALA™ ExoFit NEX™ Vest-Style Harness

Catalog No. p-4902209
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19810562 Medium
19810563 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19810562 Supplier 3M™ Supplier No. 7012816112
Only null left

Aluminum back D-ring, locking quick connect buckle leg straps, comfort padding.

  • Hybrid shoulder, back and leg comfort padding
  • Stand-up Tech-Lite™ aluminum back D-ring
  • Duo-Lok™ quick connect chest and leg straps
  • Repel™ webbing
  • Protective shoulder caps
  • Loops for body belt

Specifications

Material Repel Polyester
Color Gray
Weight (English) 4 lb.
Size Medium
Tensile Strength (English) 6000 lb.
D-Ring Location Back
Certifications/Compliance OSHA 1910.66, OSHA 1926.502, ANSI Z359.4, ANSI Z359.1, ANSI A10.32
Webbing Material Repel™ Polyester
Product Type Vest-Style Harness