missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Capillary Column End Caps

Catalog No. 03050210
Change view
Click to view available options
For Use With (Equipment):
GC Capillary Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03050210 GC Capillary Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03050210 Supplier Thermo Scientific™ Supplier No. 260EC111

Eliminate column contamination with these universal, reusable, color-coded end caps.

Eliminate column contamination with Thermo Scientific™ Capillary Column End Caps. These universal and reusable fittings are color-coded to ensure that the column is reinstalled the same way it came out.

  • Eliminate column contamination caused by leaving a column unsealed or sealed with a septum
  • Color-coordinated fittings ensure that the column is reinstalled the same way it came out
  • Reusable

Specifications

Type End Caps
Description End Caps; For Capillary Columns
For Use With (Equipment) GC Capillary Columns

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.