The Thomson Optimum Growth Integrated Lid is a sterile, vented lid specifically engineered for use with the 6Well Plate (pn931167), providing optimal gas exchange, low evaporation, and crosscontamination prevention to maintain a stable culture environment over extended growth periods. Its secure, integrated design fits precisely onto the 6Well Plate to prevent moisture loss and ingress of contaminants while simplifying handling compared to separate lids. Each lid is sterile and individually wrapped to uphold aseptic integrity during storage and setup. Thomson manufactures this product in the USA to assure sameday shipping and stringent quality control.