Learn More
Andwin Scientific Inc Cap Locks For 1.5ML Tube 100/pk DFS Item
Supplier:  Andwin Scientific Inc 16812612
The cap locks by Andwin Scientific provide reliability and security by preventing the caps of 1.5 mL tubes to pop open. This product comes in a pack of 100.
Catalog No. NC9679153
Only null left
Explore available promotions