Learn More
Brands Agilent Capillary, 0.17x90mm 1/16 in male/male
Catalog No. 50924792
Encompass
Agilent Capillary, stainless steel, 0.17 mm id, 90 mm - Standard System Capillaries, 400 and 600 bar (1100/1200/1260 Series)
UNSPSC Code  41115720
- AA
Catalog No. 50924792 Supplier Brands Supplier No. G131687300
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Agilent Capillary, stainless steel, 0.17 mm id, 90 mm - Standard System Capillaries, 400 and 600 bar (1100/1200/1260 Series)
UNSPSC Code  41115720
- AA