missing translation for 'onlineSavingsMsg'
Learn More

Research Products Corporation Cantharidin, 10 Milligrams

Catalog No. 50995818
Encompass

  • CAS Number: 56-25-7
  • Molecular Formula: C10H12O4
  • Molecular Weight: 196.2

Catalog No. 50995818 Supplier Research Products Corporation Supplier No. RPI C300700.01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • CAS Number: 56-25-7
  • Molecular Formula: C10H12O4
  • Molecular Weight: 196.2