missing translation for 'onlineSavingsMsg'
Learn More

Canrenone, 98%

Catalog No. p-7041781
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6023803 1 g
AAJ6023806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6023803 Supplier Thermo Scientific Chemicals Supplier No. J6023803
Only null left

Inhibits aldosterone biosynthesis and blocks ouabain effects | CAS: 976-71-6 | C22H28O3 | 340.463 g/mol

Inhibits aldosterone biosynthesis and blocks ouabain effects

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 976-71-6
Molecular Formula C22H28O3
Molecular Weight (g/mol) 340.463
MDL Number MFCD00071735
InChI Key UJVLDDZCTMKXJK-WNHSNXHDSA-N
Synonym canrenone, aldadiene, phanurane, luvion, aldadiene van, canrenone usan:inn, canrenonum inn-latin, canrenona inn-spanish, unii-78o20x9j0u, 17alpha-2-carboxyethyl-17beta-hydroxyandrosta-4,6-dien-3-one lactone
PubChem CID 13789
IUPAC Name (8R,9S,10R,13S,14S,17R)-10,13-dimethylspiro[2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthrene-17,5'-oxolane]-2',3-dione
SMILES CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC45CCC(=O)O5)C

Specifications

Melting Point 149°C to 151°C
Quantity 1 g
Merck Index 14,1752
Formula Weight 340.46
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Canrenone
EINECSNumber 213-554-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only