missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canrenoic acid potassium salt, 98+%
CAS: 2181-04-6 | C22H29KO4 | 396.57 g/mol
$119.16 - $1180.99
Chemical Identifiers
| CAS | 2181-04-6 |
|---|---|
| MDL Number | MFCD05662375 |
| Synonym | 3,3'-diethylthiacyanine iodide, gnf-pf-5364, 3,3'-diethylthiacyanine iodide, dye content ∼97 %, 3,3 inverted exclamation marka-diethylthiacyanine iodide, 3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2e-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2z-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6139403
|
Thermo Scientific Chemicals
J6139403 |
1 g |
Each for $119.16
|
|
|||||
|
AAJ6139406
|
Thermo Scientific Chemicals
J6139406 |
5 g |
Each for $344.23
|
|
|||||
|
AAJ6139414
|
Thermo Scientific Chemicals
J6139414 |
25 g |
Each for $1,180.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2181-04-6 | |
| MFCD05662375 | |
| 3,3'-diethylthiacyanine iodide, gnf-pf-5364, 3,3'-diethylthiacyanine iodide, dye content ∼97 %, 3,3 inverted exclamation marka-diethylthiacyanine iodide, 3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2e-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2z-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide |
Specifications
| Brown to Yellow | |
| Powder | |
| C19H19IN2S2 | |
| 14,1750 | |
| XWUVSDMFUQTKQC-UHFFFAOYSA-M | |
| (2Z)-3-ethyl-2-[(3-ethyl-1,3-benzothiazol-3-ium-2-yl)methylidene]-1,3-benzothiazole;iodide | |
| 6504364 | |
| ≥98% |
| 2181-04-6 | |
| 1 g | |
| MFCD05662375 | |
| 3,3'-diethylthiacyanine iodide, gnf-pf-5364, 3,3'-diethylthiacyanine iodide, dye content ∼97 %, 3,3 inverted exclamation marka-diethylthiacyanine iodide, 3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2e-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2z-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide | |
| CCN1C2=CC=CC=C2SC1=CC3=[N+](C4=CC=CC=C4S3)CC.[I-] | |
| 466.40 | |
| 396.58 | |
| Canrenoic acid potassium salt |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 218-554-9
RTECSNumber : TU3500000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only