missing translation for 'onlineSavingsMsg'
Learn More

Canrenoic acid potassium salt, 98+%

Catalog No. AAJ6139406
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6139406 5 g
AAJ6139403 1 g
AAJ6139414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6139406 Supplier Thermo Scientific Chemicals Supplier No. J6139406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2181-04-6 | C22H29KO4 | 396.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2181-04-6
MDL Number MFCD05662375

Specifications

Color Brown to Yellow
Physical Form Powder
Quantity 5 g
Molecular Formula C22H29KO4
Merck Index 14,1750
InChI Key JTZQCHFUGHIPDF-RYVBEKKQSA-M
SMILES CC12CCC(=O)C=C1C=CC3C2CCC4(C3CCC4(CCC(=O)[O-])O)C.[K+]
IUPAC Name potassium;3-[(10S,13R,17S)-17-hydroxy-10,13-dimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl]propanoate
Molecular Weight (g/mol) 396.57
PubChem CID 133109093
Formula Weight 396.58
Percent Purity ≥98%
Chemical Name or Material Canrenoic acid potassium salt
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 218-554-9
RTECSNumber TU3500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only