missing translation for 'onlineSavingsMsg'
Learn More

Cannon™ CT-1000 Constant-Temperature Viscosity Bath Drawer Unit

Catalog No. 1361711
Change view
Click to view available options
For Use With (Equipment):
Cannon constant temperature bath
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
1361711 Cannon constant temperature bath
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1361711 Supplier Cannon™ Supplier No. 9726A50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

For Use With (Equipment) Cannon constant temperature bath
Model CT-1000
Width (English) 18 in.
Width (Metric) 45.7 cm
Height (English) 6.5 in.
Height (Metric) 16.5 cm
Shipping Weight (Metric) 13.6 kg