missing translation for 'onlineSavingsMsg'
Learn More

Candesartan Cilexetil 98.0+%, TCI America™

Catalog No. C2635200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2635200MG 200 mg
C26351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2635200MG Supplier TCI America Supplier No. C2635200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 145040-37-5
Molecular Formula C33H34N6O6
Molecular Weight (g/mol) 610.671
MDL Number MFCD00871371
InChI Key GHOSNRCGJFBJIB-UHFFFAOYSA-N
Synonym candesartan cilexetil, atacand, amias, parapres, kenzen, candesartan cilexetil usan, candesartan cilexitil, candesartan cilextil, chembl1014
PubChem CID 2540
ChEBI CHEBI:3348
IUPAC Name 1-cyclohexyloxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
SMILES CCOC1=NC2=CC=CC(=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NNN=N5)C(=O)OC(C)OC(=O)OC6CCCCC6

Specifications

Melting Point 159°C
Color White
Quantity 200 mg
Formula Weight 610.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Candesartan Cilexetil
RTECSNumber DD6672500
TSCA No
Recommended Storage Refrigerator