missing translation for 'onlineSavingsMsg'
Learn More

Candesartan, 98%

Catalog No. p-7042521
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62818MC 100 mg
AAJ6281803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62818MC Supplier Thermo Scientific Chemicals Supplier No. J62818MC
Only null left

Angiotensin II receptor antagonist | CAS: 139481-59-7 | C24H20N6O3 | 440.463 g/mol

Candesartan is a selective AT1 (angiotensin II receptor 1) antagonist. Antagonism of angiotensin receptors inhibits vasoconstriction and the production of aldosterone, leading to a decrease in water and sodium concentration in blood plasma. Exhibits antihypertensive effects in animal models. Used in treatment of congestive heart failure, as antihypertensive. Candesartan does not affect cell viability or proliferation but increases the expression of VEGF and interleukin-8 in the cultured medium of KU-19-19 cells. Candesartan (0.1 nM) could reduce the maximal contractile response to angiostensin II by approximately 50%.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethyl acetate, methanol, water (<1mg/mL at 25°C), DMSO (88 mg/ml at 25°C), and ethanol (1 mg/ml at 25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 139481-59-7
Molecular Formula C24H20N6O3
Molecular Weight (g/mol) 440.463
MDL Number MFCD00081076
InChI Key HTQMVQVXFRQIKW-UHFFFAOYSA-N
Synonym candesartan, blopress, ratacand, 1-2'-1h-tetrazol-5-yl-1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzo d imidazole-7-carboxylic acid, candesartan ban, 3h candesartan, unii-s8q36md2xx, 2-ethoxy-3-4-2-2h-tetrazol-5-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid, candesartan usan/inn, candesartan usan:inn
PubChem CID 2541
ChEBI CHEBI:3347
IUPAC Name 2-ethoxy-3-[[4-[2-(2H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylic acid
SMILES CCOC1=NC2=CC=CC(=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C5=NNN=N5)C(=O)O

Specifications

Melting Point 183°C to 185°C
Color White
Quantity 100 mg
Merck Index 14,1739
Solubility Information Soluble in ethyl acetate,methanol,water (<1mg/ml at 25°C),DMSO (88mg/ml at 25°C),and ethanol (1mg/ml at 25°C).
Formula Weight 440.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Candesartan
Hazard Category H361d
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P281-P308+P313-P501c
RTECSNumber DD6671000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only