Learn More
Cadence Science™ Cancer Implant Needle
Catalog No. 14825284
Change view
Click to view available options
Thickness:
13 ga
Catalog No. Thickness
14825284 13 ga
1 options
Catalog No. 14825284 Supplier Cadence Science™ Supplier No. POP7927
Needle provides simple procedure to obtain and implant tumorous tissue cells beneath skin of lab animals for observation while maintaining a sterile field
  • Two piece needle set
  • Consists of sharp pointed stainless-steel outer cannula and stainless steel blunt obturator which protrudes 3.2mm beyond the needle point
Specifications
For Use With (Application) Obtain and implant tumorous tissue cells beneath the skin of lab animals
Type Cancer Implant Needle
Material Stainless Steel
Size Standard
Thickness 13 ga
Length (English) 3.25 in.
Length (Metric) 8.25 cm