missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cancer Diagnostics, Inc.™ Tissue Marking Dyes
Formulated to be used without a mordant
Supplier: Cancer Diagnostics, Inc.™ 07286
Description
- Dyes are pigment-based with an alcohol foundation for consistency and durability
- Proprietary blend of materials remains bright through processing and produces clean, crisp results
- Will not run or bleed on tissue
- Impervious to laboratory chemicals and reagents
Order Info
Individual colors available in two sizes; also available in systems with sets of six or seven colors, wood tray and 50 applicator sticks. Two-year shelf life.
Specifications
| 93°C | |
| Tissue Marking Dye | |
| Isopropyl Alcohol 400ppm, Ammonia 35ppm | |
| Liquid |
| Orange | |
| 8 oz. | |
| 8.5 to 10 |