missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cancer Diagnostics, Inc.™ Tissue Marking Dyes
Description
- Dyes are pigment-based with an alcohol foundation for consistency and durability
- Proprietary blend of materials remains bright through processing and produces clean, crisp results
- Will not run or bleed on tissue
- Impervious to laboratory chemicals and reagents
Order Info
Individual colors available in two sizes; also available in systems with sets of six or seven colors, wood tray and 50 applicator sticks. Two-year shelf life.
Specifications
Specifications
| Boiling Point | 93°C |
| Color | Orange |
| Product Type | Tissue Marking Dye |
| Quantity | 2 oz. |
| Formulation | Isopropyl Alcohol 400ppm, Ammonia 35ppm |
| pH | 8.5 to 10 |
| Form | Liquid |