missing translation for 'onlineSavingsMsg'
Learn More

Cancer Diagnostics, Inc.™ Tissue Marking Dyes

Catalog No. 22500014
Encompass
Change view
Click to view available options
Quantity:
2 oz.
8 oz.
Color:
Black
Blue
Green
Green, Black, Red, Yellow, Blue, Orange
Green, Black, Red, Yellow, Blue, Orange, Violet
Orange
Red
Yellow
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
22500014 2 oz. Green
22500017 2 oz. Orange
22500031 2 oz. Green, Black, Red, Yellow, Blue, Orange
22500466 2 oz. Green, Black, Red, Yellow, Blue, Orange, Violet
22500012 2 oz. Black
22500013 2 oz. Blue
22500015 2 oz. Red
22500019 8 oz. Blue
22500016 2 oz. Yellow
22500018 8 oz. Black
22500022 8 oz. Yellow
22500020 8 oz. Green
22500021 8 oz. Red
22500023 8 oz. Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 22500014 Supplier Cancer Diagnostics, Inc.™ Supplier No. CDI07273
Only null left

Formulated to be used without a mordant

  • Dyes are pigment-based with an alcohol foundation for consistency and durability
  • Proprietary blend of materials remains bright through processing and produces clean, crisp results
  • Will not run or bleed on tissue
  • Impervious to laboratory chemicals and reagents

Order Info

Individual colors available in two sizes; also available in systems with sets of six or seven colors, wood tray and 50 applicator sticks. Two-year shelf life.

Specifications

Boiling Point 93°C
Color Green
Product Type Tissue Marking Dye
Quantity 2 oz.
Formulation Isopropyl Alcohol 400ppm, Ammonia 35ppm
pH 8.5 to 10
Form Liquid