missing translation for 'onlineSavingsMsg'
Learn More

Canada balsam, natural filtered

Catalog No. p-7052402
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1628914 25 g
AAA1628922 100 g
AAA1628936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1628914 Supplier Thermo Scientific Chemicals Supplier No. A1628914
Only null left

Mounting medium for microscopy | CAS: 8007-47-4

Canada balsam, is used in film forming and masking. It is also used as a mounting media for microscopy. It is also used in chemical, Pharmaceutical research, pharmaceutical intermediate and research. Canada Balsam is used in the preparation of slide mounts. A turpentine made of resin which is dissolved in essential oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Canada balsam, is used in film forming and masking. It is also used as a mounting media for microscopy. It is also used in chemical, Pharmaceutical research, pharmaceutical intermediate and research. Canada Balsam is used in the preparation of slide mounts. A turpentine made of resin which is dissolved in essential oils.

Solubility
It is soluble in benzene, chloroform, xylene, ethyl acetate, ether, oil of cedar, and oil of turpentine. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air and moisture sensitive. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 8007-47-4
MDL Number MFCD00132800
Synonym Balsam Canada

Specifications

Density 0.99 g/mL
Flash Point 43°C (109°F)
Refractive Index 1.523
Quantity 25 g
UN Number UN1993
Merck Index 14,946
Solubility Information It is soluble in benzene,chloroform,xylene,ethyl acetate,ether,oil of cedar,and oil of turpentine. Insoluble in water.
Physical Form Very Viscous Liquid
Chemical Name or Material Canada balsam, Natural filtered
Hazard Category H227
Hazard Statement GHS H Statement
H226
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 232-362-2
RTECSNumber CP2352500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only