missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Canada Balsam

Catalog No. p-6911044
Change view
Click to view available options
Volume (Metric):
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
M1016910100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1016910100 Supplier MilliporeSigma™ Supplier No. 1.01691.0100
Only null left

For microscopy. Commonly used mounting medium to prepare permanent slides for microscopy. It is produced from the resin of the balsam fir tree and its use can be combined with xylene-containing specimens.

  • Density: 0.98g/cm3 (20°C)
  • CAS Registry Number: 8007-47-4

Specifications

Packaging Glass Bottle
Content And Storage 5° to 30°C
Volume (Metric) 100 mL