missing translation for 'onlineSavingsMsg'
Learn More

(+)-Camptothecin, 98%

CAS: 7689-03-4 | C20H16N2O4 | 348.35 g/mol

Supplier:  Thermo Scientific Chemicals 276720010

 View more versions of this product

Catalog No. AC276720010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(+)-Camptothecin
3-4-7689
100.0
Yellow
Authentic
Glass bottle
MFCD00081076
15, 1737
Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,acetonirile,dmso,,chloroform,hydrochloric acid
CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O
24360
348.35
Powder
98%
97.5
260.0°C
1.0% max.
97.5% min. (HPLC)
C20H16N2O4
1 g
camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecin
VSJKWCGYPAHWDS-FQEVSTJZSA-N
348.35
CHEBI:27656
98%

Chemical Identifiers

3-4-7689
348.35
VSJKWCGYPAHWDS-FQEVSTJZSA-N
24360
CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O
C20H16N2O4
MFCD00081076
camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecin
CHEBI:27656
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

RTECSNumber : UQ0492000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only