missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Camptothecin

Catalog No. AAJ62523MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62523MD 250 mg
AAJ6252303 1 g
AAJ6252306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62523MD Supplier Thermo Scientific Chemicals Supplier No. J62523MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cytotoxic antitumor agent that inhibits topoisomerase I | CAS: 7689-03-4 | C20H16N2O4

Camptothecin is used as a cytotoxic antitumor agent that inhibits topoisomerase I.Camptothecin shows remarkable anticancer activity based on preliminary clinical trials. Its analogues are used in cancer chemotherapy. Cyclodextrin-based polymer containing camptothecin is used in the investigational anti-cancer drug study.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3-4-7689
Synonym (S)-(+)-Camptothecin

Specifications

Description Yellow
Quantity 250 mg
Molecular Formula C20H16N2O4
Formula Weight 348.35g/mol
Melting Point 260°C
Product Type Apoptosis Reagent
Content And Storage 2°C to 8°C
For Use With (Application) Cytotoxic antitumor agent that inhibits topoisomerase I
Form Powder

RUO – Research Use Only