Learn More
Thermo Scientific Chemicals Camptothecin
Cytotoxic antitumor agent that inhibits topoisomerase I | CAS: 7689-03-4 | C20H16N2O4
Supplier: Thermo Scientific Chemicals J6252303
| Quantity | 1 g |
|---|
Description
Camptothecin is used as a cytotoxic antitumor agent that inhibits topoisomerase I.Camptothecin shows remarkable anticancer activity based on preliminary clinical trials. Its analogues are used in cancer chemotherapy. Cyclodextrin-based polymer containing camptothecin is used in the investigational anti-cancer drug study.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3-4-7689 | |
| (S)-(+)-Camptothecin |
Specifications
| Yellow | |
| 3-4-7689 | |
| 348.35g/mol | |
| 260°C | |
| 2°C to 8°C | |
| Powder |
| 1 g | |
| C20H16N2O4 | |
| (S)-(+)-Camptothecin | |
| Apoptosis Reagent | |
| Cytotoxic antitumor agent that inhibits topoisomerase I |