missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Camphor-10-sulfonic acid, 98%

Catalog No. AAA1262036
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1262036 500 g
AAA1262022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1262036 Supplier Thermo Scientific Chemicals Supplier No. A1262036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5872-08-2 | C10H16O4S | 232.29 g/mol

(+)-Camphor-10-sulfonic acid is used as a resolving agent for chiral amines and other cations. It is involved in the preparation of active pharmaceutical ingredient such as trimetaphan camsilate, which is used to reduce bleeding during neurosurgery. Further, it is involved in the transglucosidation of methyl and ethyl D-glucopyranosides by alcoholysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-Camphor-10-sulfonic acid is used as a resolving agent for chiral amines and other cations. It is involved in the preparation of active pharmaceutical ingredient such as trimetaphan camsilate, which is used to reduce bleeding during neurosurgery. Further, it is involved in the transglucosidation of methyl and ethyl D-glucopyranosides by alcoholysis.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 8-2-5872
Molecular Formula C10H16O4S
Molecular Weight (g/mol) 232.29
MDL Number MFCD00074827
InChI Key MIOPJNTWMNEORI-UHFFFAOYNA-N
Synonym reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid
PubChem CID 18462
ChEBI CHEBI:55379
IUPAC Name (7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C

Specifications

Melting Point ∼200°C (decomposition)
Quantity 500 g
UN Number UN3261
Beilstein 3205973
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 232.3
Percent Purity 98%
Chemical Name or Material (+/-)-Camphor-10-sulfonic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 227-527-0
RTECSNumber DT5077100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only