missing translation for 'onlineSavingsMsg'
Learn More

Camphene, 75%, remainder mainly alpha-fenchene

Catalog No. AC295570025
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295570025 2.5 L Glass bottle
AC295570010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC295570025 Supplier Thermo Scientific Chemicals Supplier No. 295570025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-92-5 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-92-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00066603
InChI Key CRPUJAZIXJMDBK-UHFFFAOYSA-N
Synonym camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229
PubChem CID 6616
ChEBI CHEBI:3830
IUPAC Name 3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
SMILES CC1(C2CCC(C2)C1=C)C

Specifications

Melting Point 42°C
Color White
Boiling Point 159.0°C to 160.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 70% min. (GC)
Packaging Glass bottle
Quantity 2.5 L
Beilstein 05, 156
Merck Index 15, 1732
Solubility Information Solubility in water: negligible. Other solubilities: soluble in ether,cyclohexane,cyclohexene,,dioxane and chloroform,slightly soluble in alcohol
Viscosity 1.6 mPa.s (50°C)
Formula Weight 136.24
Percent Purity 75%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Camphene, 75%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Flammable solid.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
If eye irritation persists: Get medical advice/attention.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiti
EINECSNumber 201-234-8
RTECSNumber EX1055000
TSCA TSCA

RUO – Research Use Only