missing translation for 'onlineSavingsMsg'
Learn More

(-)-Camphanic Acid 98.0+%, TCI America™

Catalog No. C10211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10211G 1 g
C10215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10211G Supplier TCI America Supplier No. C10211G

Chemical Identifiers

CAS 13429-83-9
Molecular Formula C10H13O4
Molecular Weight (g/mol) 197.21
MDL Number MFCD00044948,MFCD00044948,MFCD00044948
InChI Key KPWKPGFLZGMMFX-ZJUUUORDSA-M
Synonym 1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carboxylic acid, 1s---camphanic acid, 1s---camphanicacid, s---1-camphanic acid
PubChem CID 12302326
IUPAC Name (1R,4S)-4,7,7-trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carboxylate
SMILES CC1(C)[C@]2(C)CC[C@]1(OC2=O)C([O-])=O

Specifications

Melting Point 201°C
Color White-Yellow
Quantity 1 g
Formula Weight 198.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Camphanic Acid