missing translation for 'onlineSavingsMsg'
Learn More

Shield™ Campers Blanket

Catalog No. 296367460
Change view
Click to view available options
Dimensions (L x W):
60 x 80 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
296367460 60 x 80 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296367460 Supplier Shield™ Supplier No. DTC8031190

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

60 x 80 in. campers blanket

  • Gray flannel
  • Also used as stretcher blanket

Specifications

Product Type Camper/Stretcher Blanket
For Use With (Application) Stretcher Blanket or Campers Blanket
Color Gray
Material Flannel
Sterility Non-sterile
Dimensions (L x W) 60 x 80 in.