missing translation for 'onlineSavingsMsg'
Learn More

Promega cAMP-Glo™ Assay DFS Item

The cAMP-Glo™ Assay is a homogeneous, bioluminescent and high-throughput assay for measuring cAMP levels in cells

$769.29 - $4413.96

Specifications

Content And Storage -30°C to -10°C
Format 384-well Plate
For Use With (Application) For high throughput analysis of library compounds to identify modulators of GPCR activity
For Use With (Equipment) For 384-well plates
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRV1502 DFS Item
View Documents
Promega
V1502
3000 Assays
Each for $4,413.96
Only null left
 
PRV1501 DFS Item
View Documents
Promega
V1501
300 Assays
Each for $769.29
Only null left
 
Description

Description

  • Simple, Bioluminescent cAMP Detection
  • Complete in ∼45 minutes
  • Scalable to 1536-well plate formats and beyond
  • No interference by fluorescent compounds
Specifications

Specifications

-30°C to -10°C
384-well Plate
For high throughput analysis of library compounds to identify modulators of GPCR activity
For 384-well plates
Videos
SDS
Documents

Documents