missing translation for 'onlineSavingsMsg'
Learn More

Promega cAMP-Glo™ Assay

Catalog No. PRV1502
Encompass
Change view
Click to view available options
Quantity:
300 Assays
3000 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1502 3000 Assays
PRV1501 300 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV1502 Supplier Promega Supplier No. V1502

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The cAMP-Glo™ Assay is a homogeneous, bioluminescent and high-throughput assay for measuring cAMP levels in cells

  • Simple, Bioluminescent cAMP Detection
  • Complete in ∼45 minutes
  • Scalable to 1536-well plate formats and beyond
  • No interference by fluorescent compounds

Specifications

Quantity 3000 Assays
Content And Storage -30°C to -10°C
Format 384-well Plate
For Use With (Application) For high throughput analysis of library compounds to identify modulators of GPCR activity
For Use With (Equipment) For 384-well plates