missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)

For cellular and molecular biology applications

$292.48 - $292.48

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 2222.40
Product Type cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66438MCR
View Documents
Thermo Scientific Chemicals
J66438MCR
1 mg
Each for $292.48
Only null left
 
Description

Description

cAMP Dependent Protein Kinase Inhibitor (PKI 5-24) acts as inhibitor of c-AMP-dependent protein kinase These proteins bind to the catalytic subunit of cyclic-AMP dependent protein kinase, displacing the regulatory subunit and inhibiting phosphorylating activity. The cyclic-AMP binding capacity of the regulatory subunit is enhanced.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
Solid
cAMP Dependent Protein Kinase Inhibitor (PKI 5-24)
Thr-Thr-Tyr-Ala-Asp-Phe-Ile-Ala-Ser-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp
-30°C to -10°C
2222.40
RUO - research use only
Videos
SDS
Documents

Documents

RUO – Research Use Only