missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ cAMP Dependent PK Inhibitor, PKI (14-24), Amide

For cellular and molecular biology applications

Specifications

Product Type cAMP Dependent PK Inhibitor, PKI (14-24), amide
Molecular Weight (g/mol) 1251.37
Content And Storage -30°C to -10°C
Format Solid
Sequence Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp-NH2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66795MCR
View Documents
Thermo Scientific™
J66795MCR
1 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

cAMP Dependent PK Inhibitor, PKI (14-24), amide
-30°C to -10°C
Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp-NH2
HPLC
1251.37
Solid
RUO - research use only
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only