UserName
Recombinant human BUB1B (613-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.