Learn More
Cambridge Diagnostic Products, Inc.™ Camco™ Wright Stain, Fixative and Buffer
Plastic container is easy to store and allows convenient dispensing into slide stainer traps
Supplier:  Cambridge Diagnostic Products, Inc.™ 4014
 View more versions of this product
Catalog No. 04-330-4
Only null left
Explore available promotions
Description
Description
  • Contains the optimum concentration of stain to give accurate results when used on the Hemtrak™ WBC Differential System and for manual staining
  • Plastic container is easy to store and allows convenient dispensing into slide stainer traps
  • Wright stain buffer is helpful with the development of the correct, clear nuclear color schemes and has a pH of 7.15 (10 × 4.4g vial; each vial prepares 1 gallon of pH 7.15 buffer)
  • Directions for use printed on Wright stain container
Specifications
Specifications
1 gal.
Fixative and Buffer
For in-vitro diagnostic use
Product Suggestions
Product Suggestions