Learn More
Eisco™ Cam with Straight Line and Lever Followers Mechanical Demonstration Model
Catalog No. S27671
Change view
Click to view available options
Cam with Straight Line and Lever Followers Mechanical Demonstration Model
Catalog No.
S27671 Cam with Straight Line and Lever Followers Mechanical Demonstration Model
1 options
Catalog No. S27671 Supplier Eisco™ Supplier No. PH0366TM39
The Cam with Straight Line and Lever Followers is an interactive visual demonstration of the conversion of rotational motion into controlled linear motion, allowing for precise and programmable movement in mechanical systems.
  • Hand-cranked interactive model for visual learning.
  • Shows the fundamentals of how a cam with straight line and lever followers works.
  • Open system is sandwiched by two clear plastic plates for unobstructed study.
  • Constructed with heavy duty metal and plastic.
  • Includes comprehensive instruction and experiment manual.
Specifications
Product Type Cam with Straight Line and Lever Followers Mechanical Demonstration Model