Learn More
MP Biomedicals™ Cam Lock
Cam Lock is a locking system for FastPrep-24™ 5G Instruments.
Supplier: MP Biomedicals™ 116005588
Description
Modeled after the bike tire quick release, the familiar mechanism allows:- Rapid sample exchange
- Strong grip
- Uniform clamping force
- Ease of use
Specifications
| FastPrep-24™ 5G Instruments | |
| 1 Pk. |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.