missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Ether, Stabilized, ACS, 99%, Spectrum™ Chemical

Catalog No. 18611145
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611145 200 L Amber Glass Bottle
18611141 250 mL Amber Glass Bottle
18611142 1 L Amber Glass Bottle
18611143 2.5 L Amber Glass Bottle
18611144 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611145 Supplier Spectrum Chemical Mfg Cor Supplier No. I1052200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-20-3

Specifications

CAS Max % 1
Boiling Point 68°C (154.4°F)
Packaging Amber Glass Bottle
Quantity 200 L
Residue after Evaporation 0.0001
Percent Purity 0.99
Grade ACS
Titratable Acid 0.0007 meq/g
Physical Form Liquid
Chemical Name or Material Isopropyl Ether, Stabilized

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.