missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calphostin C, 95%

Catalog No. AAJ60647EXG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60647EXG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60647EXG Supplier Thermo Scientific Chemicals Supplier No. J60647EXG

 

Potent inhibitor of protein kinase C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Purification Method HPLC
Purity 95%
Format Powder
For Use With (Application) Potent inhibitor of protein kinase C
Molecular Weight (g/mol) 790.76
Product Type Calphostin C, 95%
Quantity 100 μg
Regulatory Status RUO
Sequence UCN-1028c

RUO – Research Use Only