missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calpain Inhibitor III, 95+%

CAS: 88191-84-8 | C22H26N2O4, C19H35N3O4S | 382.46 g/mol

Supplier:  Thermo Scientific Chemicals J62919MB

Catalog No. AAJ62919MB


Only null left

Description

Description

Inhibits calpain proteolysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

25 mg
2°C to 8°C
≥95%
C22H26N2O4
MDL; Z-Val-Phe-CHO
Signal Transduction Reagent
NGBKFLTYGSREKK-PMACEKPBSA-N
benzyl N-[(1S)-2-methyl-1-{[(2S)-1-oxo-3-phenylpropan-2-yl]carbamoyl}propyl]carbamate
White
Inhibits calpain proteolysis
88191-84-8
401.60g/mol
Soluble in DMSO at 25mg/ml
Powder
CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)N[C@@H](CC1=CC=CC=C1)C=O
382.46

Chemical Identifiers

88191-84-8
382.46
MDL; Z-Val-Phe-CHO
CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)N[C@@H](CC1=CC=CC=C1)C=O
C22H26N2O4
NGBKFLTYGSREKK-PMACEKPBSA-N
benzyl N-[(1S)-2-methyl-1-{[(2S)-1-oxo-3-phenylpropan-2-yl]carbamoyl}propyl]carbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 618-124-4

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only