missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calpain Inhibitor I, 95+%

Catalog No. AAJ61766LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61766LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61766LB0 Supplier Thermo Scientific Chemicals Supplier No. J61766LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

It inhibits apoptosis in thymocytes and metamyelocytes. It also inhibits cell cycle progression at the G1/S border and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. ALLN inhibits the proteolysis of IκBα and IκBβ by the ubiquitin-proteasome complex. ALLN also prevents nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110044-82-1
Molecular Formula C20H37N3O4
Synonym Ac-LLnL-CHO; ALLN

Specifications

Quantity 5 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) An inhibitor of both Calpain I and II
Formula Weight 383.53
Form Powder
Assay 95+%

RUO – Research Use Only