Learn More
DWK Life Sciences Calibrex™ solutae 530 Bottle Top Dispenser with Flow Control Stopcock
Fits most laboratory bottles either directly or using supplied adapters. Calibrex™ solutae 530 Bottle Top Dispensers with Flow Control Stopcock feature excellent chemical resistance, high volumetry performance and simplified maintenance. Volume rang
$3,766.25
Specifications
| Product Type | Bottletop Dispenser |
|---|---|
| Autoclavable | Autoclavable |
| For Use With (Equipment) | Fits most laboratory bottles directly or with supplied adapters |
| Includes | Flow Control Stopcock, PFA-coated plunger and integrated calibration |
| Model | Calibrex solutae 530 |
Description
The Calibrex™ 530 solutae has a PFA-coated plunger preventing the plunger and barrel from seizing together. This enables trouble free distribution of salt solutions, weak and strong acids, as well as bases. The materials in contact with liquid flow are chemically inert, providing for stability and long instrument life.
- Flow control stopcock enables liquid priming and recycling without reagent loss or contamination
- Permanent fluid path visibility
- Integrated calibration system
- Choice of adjustment button with each unit
- Long lasting performance stability
- Autoclavable at 121°C / 250°F fully assembled
- Two year warranty
Specifications
| Bottletop Dispenser | |
| Fits most laboratory bottles directly or with supplied adapters | |
| Calibrex solutae 530 |
| Autoclavable | |
| Flow Control Stopcock, PFA-coated plunger and integrated calibration | |
| 2 Years |